6-Bromo-1-methyl-1H-pyrrolo[3,2-c]pyridine 95%
Supplier: Ambeed
|
Warning
|
A507927-100MG
A507927-250MG
A507927-1G
76649-462EA
54.11
USD
76649-462
76649-464
77749-037
6-Bromo-1-methyl-1H-pyrrolo[3,2-c]pyridine 95%
6-Bromo-1-methyl-1H-pyrrolo[3,2-c]pyridine
Precaución: for Research Use Only
Formula:
C₈H₇BrN₂ MW: 211.0586 g/mol |
MDL Number:
MFCD30067319 CAS Number: 2091221-98-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...