Tris(3-hydroxypropyltriazolylmethyl)amine 97%

Supplier: Ambeed
Warning

Synonyms: THPTA, Tris[[1-(3-hydroxypropyl)-1H-1,2,3-triazol-4-yl]methyl]amine, 3,3′,3′′-(4,4′,4′′-(Nitrilotris(methylene))tris(1H-1,2,3-triazole-4,1-diyl))tris(propan-1-ol)

A506578-100MG A506578-1G A506578-250MG A506578-5G
76649-446EA 65.78 USD
76649-446 76649-448 76649-450 76649-452
Tris(3-hydroxypropyltriazolylmethyl)amine 97%
Tris(3-hydroxypropyltriazolylmethyl)amine

Precaución: for Research Use Only

Formula: C₁₈H₃₀N₁₀O₃
MW: 434.51 g/mol
MDL Number: MFCD27665386
CAS Number: 760952-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR