7-Methylnaphthalen-2-ol 95%

Supplier: Ambeed
Warning

A505632-100MG A505632-250MG A505632-1G
76649-442EA 48.81 USD
76649-442 76649-444 77749-031
7-Methylnaphthalen-2-ol 95%
7-Methylnaphthalen-2-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₀O
MW: 158.2 g/mol
MDL Number: MFCD09037748
CAS Number: 26593-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR