Ethyl 5-amino-1H-imidazole-4-carboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A504306-250MG A504306-5G
76649-430EA 79.57 USD
76649-430 76649-432
Ethyl 5-amino-1H-imidazole-4-carboxylate hydrochloride 95%
Ethyl 5-amino-1H-imidazole-4-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₀ClN₃O₂
MW: 191.62 g/mol
MDL Number: MFCD06245145
CAS Number: 118019-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR