4,7-Dibromo-1H-benzo[d]imidazole 98%

Supplier: Ambeed
Warning

A503788-1G A503788-250MG A503788-5G
76649-424EA 43 USD
76649-424 76649-426 76649-428
4,7-Dibromo-1H-benzo[d]imidazole 98%
4,7-Dibromo-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₇H₄Br₂N₂
MW: 275.9281 g/mol
MDL Number: MFCD16872346
CAS Number: 148185-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR