Methyl 5-hydroxypentanoate 95%

Supplier: Ambeed
Warning

A499937-1G A499937-5G A499937-250MG
76649-412EA 49 USD
76649-412 76649-414 77598-522
Methyl 5-hydroxypentanoate 95%
Methyl 5-hydroxypentanoate

Precaución: for Research Use Only

Formula: C₆H₁₂O₃
MDL Number: MFCD13689062
CAS Number: 14273-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR