tert-Butyl 3-(bromomethyl)bicyclo[1.1.1]pentane-1-carboxylate 98%

Supplier: Ambeed
Warning

A495292-250MG A495292-100MG
76649-386EA 960.12 USD
76649-386 77749-014
tert-Butyl 3-(bromomethyl)bicyclo[1.1.1]pentane-1-carboxylate 98%
tert-Butyl 3-(bromomethyl)bicyclo[1.1.1]pentane-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₇BrO₂
MW: 261.155 g/mol
MDL Number: MFCD29037328
CAS Number: 138732-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR