2-(3-(Cyclopropylmethoxy)-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%

Supplier: Ambeed
Warning

A494282-100MG A494282-1G A494282-250MG
76649-372EA 37.14 USD
76649-372 76649-374 76649-376
2-(3-(Cyclopropylmethoxy)-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%
2-(3-(Cyclopropylmethoxy)-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₇H₂₂BF₃O₃
MW: 342.1610 g/mol
MDL Number: MFCD18434456
CAS Number: 2096334-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR