5-(4-(Isopropylthio)phenyl)-1H-pyrrole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A490145-100MG A490145-250MG
76649-366EA 125.19 USD
76649-366 76649-368
5-(4-(Isopropylthio)phenyl)-1H-pyrrole-2-carboxylic acid 97%
5-(4-(Isopropylthio)phenyl)-1H-pyrrole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO₂S
MW: 261.3400 g/mol
CAS Number: 1538665-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR