1-(6-Amino-5-methylpyridin-3-yl)ethanone 97%

Supplier: Ambeed
Warning

A489533-100MG A489533-1G A489533-250MG
76649-360EA 134.74 USD
76649-360 76649-362 76649-364
1-(6-Amino-5-methylpyridin-3-yl)ethanone 97%
1-(6-Amino-5-methylpyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
MDL Number: MFCD11100589
CAS Number: 1033202-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR