(4R,5S)-4,5-Diphenyl-2-(5-(trifluoromethyl)pyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1349592-100MG A1349592-1G A1349592-250MG
76658-094EA 45 USD
76658-094 76658-096 76658-098
(4R,5S)-4,5-Diphenyl-2-(5-(trifluoromethyl)pyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%
(4R,5S)-4,5-Diphenyl-2-(5-(trifluoromethyl)pyridin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₁H₁₅F₃N₂O
MW: 368.3518 g/mol
CAS Number: 2635322-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR