(E)-N'-(1-(5-Chloro-2-hydroxyphenyl)ethylidene)-3-((4-methylpiperazin-1-yl)sulfonyl)benzohydrazide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Seclidemstat
A486013-10MG
A486013-1MG
A486013-25MG
A486013-5MG
A486013-50MG
76649-344EA
47.75
USD
76649-344
76649-346
76649-348
76649-350
77622-338
(E)-N'-(1-(5-Chloro-2-hydroxyphenyl)ethylidene)-3-((4-methylpiperazin-1-yl)sulfonyl)benzohydrazide 97%
(E)-N'-(1-(5-Chloro-2-hydroxyphenyl)ethylidene)-3-((4-methylpiperazin-1-yl)sulfonyl)benzohydrazide
Precaución: for Research Use Only
Formula:
C₂₀H₂₃ClN₄O₄S MW: 450.94 g/mol |
MDL Number:
MFCD31630770 CAS Number: 1423715-37-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...