3-Chloro-4-cyanopicolinic acid 95%

Supplier: Ambeed
Warning

A485094-100MG A485094-1G A485094-250MG
76649-338EA 41 USD
76649-338 76649-340 76649-342
3-Chloro-4-cyanopicolinic acid 95%
3-Chloro-4-cyanopicolinic acid

Precaución: for Research Use Only

Formula: C₇H₃ClN₂O₂
MDL Number: MFCD28749706
CAS Number: 1805153-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR