3,5-Dibromo-1H-pyrazole 98%

Supplier: Ambeed
Warning

A484983-100G A484983-1G A484983-250MG A484983-25G A484983-5G
76649-328EA 490.14 USD
76649-328 76649-330 76649-332 76649-334 76649-336
3,5-Dibromo-1H-pyrazole 98%
3,5-Dibromo-1H-pyrazole

Precaución: for Research Use Only

Formula: C₃H₂Br₂N₂
MW: 225.869 g/mol
MDL Number: MFCD00159645
CAS Number: 67460-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR