Benzyl (S)-4-(4-amino-6,7-dimethoxyquinazolin-2-yl)-2-(tert-butylcarbamoyl)piperazine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: L-765314

A484697-10MG A484697-1MG A484697-25MG A484697-50MG A484697-5MG
76649-312EA 175.05 USD
76649-312 76649-314 76649-316 76649-318 76649-320
Benzyl (S)-4-(4-amino-6,7-dimethoxyquinazolin-2-yl)-2-(tert-butylcarbamoyl)piperazine-1-carboxylate 97%
Benzyl (S)-4-(4-amino-6,7-dimethoxyquinazolin-2-yl)-2-(tert-butylcarbamoyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₂₇H₃₄N₆O₅
MW: 522.5961 g/mol
MDL Number: MFCD05664727
CAS Number: 189349-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR