(6-Chloro-2-fluoropyridin-3-yl)methanol 95%

Supplier: Ambeed
Warning

A483840-100MG A483840-1G A483840-250MG A483840-5G
76649-294EA 25 USD
76649-294 76649-296 76649-298 76649-300
(6-Chloro-2-fluoropyridin-3-yl)methanol 95%
(6-Chloro-2-fluoropyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₅ClFNO
MDL Number: MFCD16610401
CAS Number: 1227509-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR