5-(tert-Butyl)-1H-benzo[d]imidazole 95%

Supplier: Ambeed
Warning

A483618-100MG A483618-1G A483618-250MG A483618-5G
76649-286EA 19 USD
76649-286 76649-288 76649-290 76649-292
5-(tert-Butyl)-1H-benzo[d]imidazole 95%
5-(tert-Butyl)-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂
MDL Number: MFCD19373985
CAS Number: 110534-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR