(S)-N-(3-(6-Isopropoxypyridin-3-yl)-1H-indazol-5-yl)-1-(2-(4-(4-(1-methyl-1H-1,2,4-triazol-3-yl)phenyl)-3,6-dihydropyridin-1(2H)-yl)-2-oxoethyl)-3-(methylthio)pyrrolidine-3-carboxamide 95%

Supplier: Ambeed
Warning

Synonyms: MK-8353

A480964-2MG A480964-5MG A480964-1MG
76649-272EA 23.34 USD
76649-272 76649-274 77622-334
(S)-N-(3-(6-Isopropoxypyridin-3-yl)-1H-indazol-5-yl)-1-(2-(4-(4-(1-methyl-1H-1,2,4-triazol-3-yl)phenyl)-3,6-dihydropyridin-1(2H)-yl)-2-oxoethyl)-3-(methylthio)pyrrolidine-3-carboxamide 95%
(S)-N-(3-(6-Isopropoxypyridin-3-yl)-1H-indazol-5-yl)-1-(2-(4-(4-(1-methyl-1H-1,2,4-triazol-3-yl)phenyl)-3,6-dihydropyridin-1(2H)-yl)-2-oxoethyl)-3-(methylthio)pyrrolidine-3-carboxamide

Precaución: for Research Use Only

Formula: C₃₇H₄₁N₉O₃S
MW: 691.85 g/mol
CAS Number: 1184173-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR