3-(Methoxymethyl)cyclobutane-1-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A479665-100MG
A479665-250MG
76649-266EA
337.37
USD
76649-266
76649-268
3-(Methoxymethyl)cyclobutane-1-carboxylic acid 97%
3-(Methoxymethyl)cyclobutane-1-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₁₂O₃ MW: 144.1700 g/mol |
CAS Number:
408326-43-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...