Boc-​Val-​Cit-​PABC-​PNP 98%

Supplier: Ambeed
Warning

Synonyms: Boc-Val-Cit-PAB-PNP, tert-Butyl ((S)-3-methyl-1-(((S)-1-((4-((((4-nitrophenoxy)carbonyl)oxy)methyl)phenyl)amino)-1-oxo-5-ureidopentan-2-yl)amino)-1-oxobutan-2-yl)carbamate

A479352-250MG A479352-100MG A479352-10MG A479352-5MG
76649-264EA 250 USD
76649-264 77617-072 77748-993 77748-994
Boc-​Val-​Cit-​PABC-​PNP 98%
Boc-​Val-​Cit-​PABC-​PNP

Precaución: for Research Use Only

Formula: C₃₀H₄₀N₆O₁₀
MW: 644.7 g/mol
MDL Number: MFCD22417105
CAS Number: 870487-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR