3-Oxa-6-azabicyclo[3.1.1]heptane 4-methylbenzene-1-sulfonic acid 97%

Supplier: Ambeed
Warning

A478113-100MG A478113-250MG A478113-50MG
76649-256EA 192.03 USD
76649-256 76649-258 76649-260
3-Oxa-6-azabicyclo[3.1.1]heptane 4-methylbenzene-1-sulfonic acid 97%
3-Oxa-6-azabicyclo[3.1.1]heptane 4-methylbenzene-1-sulfonic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₄S
MW: 271.33 g/mol
MDL Number: MFCD28383857
CAS Number: 1414860-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR