8-Bromo-3-fluoroquinolin-2(1H)-one 95%

Supplier: Ambeed
Warning

A477799-100MG A477799-250MG
76649-252EA 174 USD
76649-252 76649-254
8-Bromo-3-fluoroquinolin-2(1H)-one 95%
8-Bromo-3-fluoroquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₉H₅BrFNO
MDL Number: MFCD30177936
CAS Number: 834883-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR