5,6-Dichloropyrimidin-4-ol 95%

Supplier: Ambeed
Warning

Synonyms: 5,6-Dichloropyrimidin-4(3H)-one

A477162-100MG A477162-1G A477162-250MG A477162-5G
76649-236EA 13 USD
76649-236 76649-238 76649-240 76649-242
5,6-Dichloropyrimidin-4-ol 95%
5,6-Dichloropyrimidin-4-ol

Precaución: for Research Use Only

Formula: C₄H₂Cl₂N₂O
MW: 164.9775 g/mol
MDL Number: MFCD24393897
CAS Number: 88982-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR