(2R,4R)-4-Aminopyrrolidine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A476515-250MG A476515-100MG
76649-234EA 224.92 USD
76649-234 77748-988
(2R,4R)-4-Aminopyrrolidine-2-carboxylic acid 95%
(2R,4R)-4-Aminopyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₁₀N₂O₂
MW: 130.1451 g/mol
MDL Number: MFCD08704532
CAS Number: 1217598-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR