5,6,7,8-Tetrahydroimidazo[1,5-a]pyridin-7-amine dihydrochloride 97%

Supplier: Ambeed
Warning

A475520-100MG A475520-1G A475520-250MG A475520-50MG
76649-220EA 176.11 USD
76649-220 76649-222 76649-224 76649-226
5,6,7,8-Tetrahydroimidazo[1,5-a]pyridin-7-amine dihydrochloride 97%
5,6,7,8-Tetrahydroimidazo[1,5-a]pyridin-7-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₃Cl₂N₃
MW: 210.1042 g/mol
MDL Number: MFCD26593755
CAS Number: 1417635-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR