2-(Undecyloxy)ethanol 98%

Supplier: Ambeed
Warning

A475477-1G A475477-250MG A475477-5G A475477-25G
76649-214EA 40 USD
76649-214 76649-216 76649-218 77783-429
2-(Undecyloxy)ethanol 98%
2-(Undecyloxy)ethanol

Precaución: for Research Use Only

Formula: C₁₃H₂₈O₂
MDL Number: MFCD06252492
CAS Number: 38471-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR