(S)-(-)-3-(tert-Butoxycarbonyl)-4-formyl-2,2-dimethyl-1,3-oxazolidine 95%

Supplier: Ambeed
Warning

Synonyms: (S)-(-)-3-Boc-4-formyl-2,2-dimethyl-1,3-oxazolidine, (S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde, tert-Butyl (S)-(-)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, (S)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde

A475462-10G A475462-250MG A475462-25G A475462-1G A475462-5G
76649-210EA 235.52 USD
76649-210 76649-212 77551-450 77568-936 77598-510
(S)-(-)-3-(tert-Butoxycarbonyl)-4-formyl-2,2-dimethyl-1,3-oxazolidine 95%
(S)-(-)-3-(tert-Butoxycarbonyl)-4-formyl-2,2-dimethyl-1,3-oxazolidine

Precaución: For research use only.

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
MDL Number: MFCD00209557
CAS Number: 102308-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR