FX1 97%

Supplier: Ambeed
Warning

Synonyms: (5Z)-5-(5-Chloro-1,2-dihydro-2-oxo-3H-indol-3-ylidene)-4-oxo-2-thioxo-3-thiazolidinepropanoic acid, (Z)-3-(5-(5-Chloro-2-oxoindolin-3-ylidene)-4-oxo-2-thioxothiazolidin-3-yl)propanoic acid

A475221-10MG A475221-1G A475221-1MG A475221-25MG A475221-5MG
76649-200EA 119.89 USD
76649-200 76649-202 76649-204 76649-206 76649-208
FX1 97%
FX1

Precaución: for Research Use Only

Formula: C₁₄H₉ClN₂O₄S₂
MW: 368.81 g/mol
MDL Number: MFCD31560474
CAS Number: 1426138-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR