Methyl 4-amino-6-chloropyridine-2-carboxylate 94%

Supplier: Ambeed
Warning

A473587-100MG A473587-1G A473587-250MG
76649-182EA 116.7 USD
76649-182 76649-184 76649-186
Methyl 4-amino-6-chloropyridine-2-carboxylate 94%
Methyl 4-amino-6-chloropyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₂
MW: 186.5957 g/mol
MDL Number: MFCD18206770
CAS Number: 1235475-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR