Isobutyl methanesulfonate 98% (by GC)

Supplier: Ambeed
Warning

Synonyms: Methanesulphonic acid Isobutyl ester

A469601-1G A469601-250MG A469601-5G
76649-176EA 57.29 USD
76649-176 76649-178 76649-180
Isobutyl methanesulfonate 98% (by GC)
Isobutyl methanesulfonate

Precaución: for Research Use Only

Formula: C₅H₁₂O₃S
MW: 152.21 g/mol
MDL Number: MFCD07363982
CAS Number: 16156-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR