6-Chloro-1H-pyrazolo[3,4-b]pyridine-3-carbonitrile 98%
Supplier: Ambeed
|
Warning
|
A469434-100MG
A469434-1G
A469434-250MG
76649-170EA
200.52
USD
76649-170
76649-172
76649-174
6-Chloro-1H-pyrazolo[3,4-b]pyridine-3-carbonitrile 98%
6-Chloro-1H-pyrazolo[3,4-b]pyridine-3-carbonitrile
Precaución: for Research Use Only
Formula:
C₇H₃ClN₄ MW: 178.5785 g/mol |
MDL Number:
MFCD26516651 CAS Number: 1935287-10-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...