2-Hexyldecan-1-amine ≥98% (by GC, titration analysis)

Supplier: Ambeed
Danger

A469168-100MG A469168-250MG A469168-5G A469168-25G A469168-1G
76649-164EA 7.43 USD
76649-164 76649-166 76649-168 77568-926 77617-036
2-Hexyldecan-1-amine ≥98% (by GC, titration analysis)
2-Hexyldecan-1-amine

Precaución: For research use only.

Formula: C₁₆H₃₅N
MW: 241.46 g/mol
MDL Number: MFCD30475384
CAS Number: 62281-05-4
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR