Polyethylene glycol 600 ∼ Mn 600

Supplier: Ambeed
Warning

Synonyms: Ethylene glycol polymer average Mn 600, PEG 600

A466629-1000G A466629-100G A466629-500G
76649-134EA 15.92 USD
76649-134 77598-500 77598-502
Polyethylene glycol 600 ∼ Mn 600
Polyethylene glycol 600

Precaución: for Research Use Only

Formula: H(OCH₂CH₂)nOH
MW: 600 g/mol
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR