Methyl 5-chloro-3-(chlorosulfonyl)thiophene-2-carboxylate 95%

Supplier: Ambeed
Danger

A466606-100G A466606-1G A466606-5G A466606-25G
76649-128EA 178 USD
76649-128 76649-130 76649-132 77551-418
Methyl 5-chloro-3-(chlorosulfonyl)thiophene-2-carboxylate 95%
Methyl 5-chloro-3-(chlorosulfonyl)thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₄Cl₂O₄S₂
MW: 275.1300 g/mol
MDL Number: MFCD09032981
CAS Number: 126910-68-7
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR