4-Acetyl-2-hydroxybenzoic acid 95%

Supplier: Ambeed
Warning

A466072-100MG A466072-250MG
76649-124EA 99.73 USD
76649-124 77748-951
4-Acetyl-2-hydroxybenzoic acid 95%
4-Acetyl-2-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈O₄
MW: 180.1574 g/mol
MDL Number: MFCD20639625
CAS Number: 107806-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR