3-(1,3-Dioxolan-2-yl)aniline 95%

Supplier: Ambeed
Warning

A465695-250MG A465695-5G
76649-122EA 18.04 USD
76649-122 77748-950
3-(1,3-Dioxolan-2-yl)aniline 95%
3-(1,3-Dioxolan-2-yl)aniline

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MDL Number: MFCD00272236
CAS Number: 6398-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR