N-Ethyl-4-(2-(4-fluoro-2,6-dimethylphenoxy)-5-(2-hydroxypropan-2-yl)phenyl)-6-methyl-7-oxo-6,7-dihydro-1H-pyrrolo[2,3-c]pyridine-2-carboxamide ≥99%

Supplier: Ambeed
Warning

Synonyms: ABBV-744

A464519-10MG A464519-1MG A464519-25MG A464519-5MG A464519-100MG A464519-250MG A464519-50MG
76649-112EA 109.28 USD
76649-112 76649-114 76649-116 77598-498 77622-320 77622-322 77622-324
N-Ethyl-4-(2-(4-fluoro-2,6-dimethylphenoxy)-5-(2-hydroxypropan-2-yl)phenyl)-6-methyl-7-oxo-6,7-dihydro-1H-pyrrolo[2,3-c]pyridine-2-carboxamide ≥99%
N-Ethyl-4-(2-(4-fluoro-2,6-dimethylphenoxy)-5-(2-hydroxypropan-2-yl)phenyl)-6-methyl-7-oxo-6,7-dihydro-1H-pyrrolo[2,3-c]pyridine-2-carboxamide

Precaución: for Research Use Only

Formula: C₂₈H₃₀FN₃O₄
MW: 491.55 g/mol
MDL Number: MFCD31657409
CAS Number: 2138861-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR