2-Hydroxy-3-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Warning

Synonyms: 3-(Trifluoromethyl)pyridin-2-ol

A453429-25G A453429-5G
76649-088EA 14.86 USD
76649-088 76649-090
2-Hydroxy-3-(trifluoromethyl)pyridine 95%
2-Hydroxy-3-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₄F₃NO
MW: 163.1 g/mol
MDL Number: MFCD00153194
CAS Number: 22245-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR