((3-Bromophenyl)ethynyl)trimethylsilane 95%

Supplier: Ambeed
Warning

A452899-100MG A452899-1G A452899-250MG
76649-078EA 10.61 USD
76649-078 76649-080 76649-082
((3-Bromophenyl)ethynyl)trimethylsilane 95%
((3-Bromophenyl)ethynyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrSi
MW: 253.2104 g/mol
MDL Number: MFCD01863664
CAS Number: 3989-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR