5-Fluoro-6-methyl-1H-benzo[d][1,2,3]triazole 95%
Supplier: Ambeed
|
Warning
|
A452211-100MG
A452211-1G
A452211-250MG
76649-072EA
34
USD
76649-072
76649-074
76649-076
5-Fluoro-6-methyl-1H-benzo[d][1,2,3]triazole 95%
5-Fluoro-6-methyl-1H-benzo[d][1,2,3]triazole
Precaución: for Research Use Only
Formula:
C₇H₆FN₃ |
MDL Number:
MFCD31699216 CAS Number: 2384975-27-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...