Heptadecan-9-amine 96%

Supplier: Ambeed
Danger

A450838-25G A450838-5G A450838-1G
76649-068EA 345 USD
76649-068 76649-070 77622-310
Heptadecan-9-amine 96%
Heptadecan-9-amine

Precaución: for Research Use Only

Formula: C₁₇H₃₇N
MDL Number: MFCD30742905
CAS Number: 3241-20-1
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR