2-(Oxetan-2-yl)acetic acid 98%

Supplier: Ambeed
Warning

A449485-250MG A449485-100MG
76649-064EA 333.13 USD
76649-064 77748-913
2-(Oxetan-2-yl)acetic acid 98%
2-(Oxetan-2-yl)acetic acid

Precaución: for Research Use Only

Formula: C₅H₈O₃
MW: 116.12 g/mol
MDL Number: MFCD26959068
CAS Number: 1416271-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR