4-Chloro-3-nitrobenzene-1,2-diamine 95%
Supplier: Ambeed
|
Warning
|
A446217-100MG
A446217-250MG
A446217-50MG
76649-038EA
84.88
USD
76649-038
76649-040
76649-042
4-Chloro-3-nitrobenzene-1,2-diamine 95%
4-Chloro-3-nitrobenzene-1,2-diamine
Precaución: for Research Use Only
Formula:
C₆H₆ClN₃O₂ MW: 187.5837 g/mol |
MDL Number:
MFCD02169899 CAS Number: 144729-44-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...