Methyl [2,3'-bipyridine]-6'-carboxylate 95%

Supplier: Ambeed
Warning

A442757-100MG A442757-250MG
76649-028EA 30.77 USD
76649-028 76649-030
Methyl [2,3'-bipyridine]-6'-carboxylate 95%
Methyl [2,3'-bipyridine]-6'-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O₂
MW: 214.2200 g/mol
MDL Number: MFCD29762323
CAS Number: 845827-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR