3-(tert-Butyl)-6-chloropicolinonitrile 95+%

Supplier: Ambeed
Danger

A441286-100MG A441286-1G A441286-250MG
76649-022EA 217.49 USD
76649-022 76649-024 76649-026
3-(tert-Butyl)-6-chloropicolinonitrile 95+%
3-(tert-Butyl)-6-chloropicolinonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClN₂
MW: 194.6607 g/mol
MDL Number: MFCD31698238
CAS Number: 2090123-47-8
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR