6-Methoxy-4H-chromen-4-one 97%

Supplier: Ambeed
Warning

A441268-250MG A441268-1G
76649-020EA 10.61 USD
76649-020 77616-896
6-Methoxy-4H-chromen-4-one 97%
6-Methoxy-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₁₀H₈O₃
MW: 176.1687 g/mol
MDL Number: MFCD02089718
CAS Number: 59887-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR