Potassium (cyclobutylmethyl)trifluoroborate 95%
Supplier: Ambeed
|
Warning
|
A440649-250MG
A440649-1G
A440649-100MG
76649-010EA
23
USD
76649-010
77748-900
77748-899
Potassium (cyclobutylmethyl)trifluoroborate 95%
Potassium (cyclobutylmethyl)trifluoroborate
Precaución: for Research Use Only
Formula:
C₅H₉BF₃K MW: 176.0295 g/mol |
MDL Number:
MFCD11052657 CAS Number: 2135480-21-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...