(1-Benzyl-1H-indol-5-yl)boronic acid 96%

Supplier: Ambeed
Warning

A440581-100MG A440581-1G A440581-250MG
76649-004EA 39 USD
76649-004 76649-006 76649-008
(1-Benzyl-1H-indol-5-yl)boronic acid 96%
(1-Benzyl-1H-indol-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₄BNO₂
MDL Number: MFCD30182379
CAS Number: 2346512-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR