2-Bromo-1-(3,3-difluorocyclobutyl)ethanone 98%
Supplier: Ambeed
|
Danger
|
A439481-250MG
A439481-50MG
76649-000EA
323.58
USD
76649-000
76649-002
2-Bromo-1-(3,3-difluorocyclobutyl)ethanone 98%
2-Bromo-1-(3,3-difluorocyclobutyl)ethanone
Precaución: for Research Use Only
Formula:
C₆H₇BrF₂O MW: 213.02 g/mol |
MDL Number:
MFCD28501703 CAS Number: 1619911-80-6 UN: 3265 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...