Benzyl 3,8-diazabicyclo[3.2.1]octane-8-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A438468-100MG
A438468-250MG
76648-988EA
108.22
USD
76648-988
76648-990
Benzyl 3,8-diazabicyclo[3.2.1]octane-8-carboxylate 97%
Benzyl 3,8-diazabicyclo[3.2.1]octane-8-carboxylate
Precaución: for Research Use Only
Formula:
C₁₄H₁₈N₂O₂ MW: 246.3049 g/mol |
MDL Number:
MFCD26516142 CAS Number: 956794-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...